Assay Details
| Gene Symbol: OST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342244.1 | NP_001320129.1 | 4 - 5 | 843 | 119 | |
| NM_119556.3 | NP_567945.1 | 4 - 5 | 1015 | 119 | ||
| GenBank mRNA | AY054624.1 | - | 4 - 5 | 441 | 119 | |
| AY081538.1 | - | 4 - 5 | 396 | 119 | ||
More Information
Set Membership: |
|

OST1
Genomic Map