Assay Details
| Gene Symbol: OST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119556.3 | NP_567945.1 | 9 - 10 | 1614 | 81 | |
| GenBank mRNA | AY054624.1 | - | 9 - 10 | 1040 | 81 | |
| AY081538.1 | - | 9 - 10 | 995 | 81 | ||
More Information
Set Membership: |
|

OST1
Genomic Map