Assay Details
| Gene Symbol: MBR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119565.4 | NP_195130.1 | 3 - 4 | 2176 | 84 | |
| GenBank mRNA | AK229265.1 | - | 3 - 4 | 1978 | 84 | |
| BT015366.1 | - | 3 - 4 | 1881 | 84 | ||
| BT020341.1 | - | 1 - 2 | 1576 | 84 | ||
More Information
Set Membership: |
|

MBR2
Genomic Map