Assay Details
| Gene Symbol: MBR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119565.4 | NP_195130.1 | 6 - 7 | 2466 | 79 | |
| GenBank mRNA | AK229265.1 | - | 6 - 7 | 2268 | 79 | |
| BT015366.1 | - | 6 - 7 | 2171 | 79 | ||
| BT020341.1 | - | 4 - 5 | 1866 | 79 | ||
More Information
Set Membership: |
|

MBR2
Genomic Map