Assay Details
| Gene Symbol: PAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119572.7 | NP_195137.5 | 4 - 5 | 1561 | 67 | |
| GenBank mRNA | BT002354.1 | - | 4 - 5 | 1120 | 67 | |
| L19418.1 | - | 4 - 5 | 1451 | 67 | ||
More Information
Set Membership: |
|

PAB2
Genomic Map