Assay Details
| Gene Symbol: CAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036711.1 | NP_001031788.1 | 2 - 3 | 257 | 92 | |
| NM_119587.4 | NP_195149.1 | 2 - 3 | 478 | 92 | ||
| GenBank mRNA | AY034919.1 | - | 2 - 3 | 257 | 92 | |
| AY113919.1 | - | 2 - 3 | 204 | 92 | ||
| AY302082.1 | - | 2 - 3 | 204 | 92 | ||
More Information
Set Membership: |
|

CAD5
Genomic Map