Assay Details
| Gene Symbol: CAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036711.1 | NP_001031788.1 | 3 - 4 | 483 | 76 | |
| NM_119587.4 | NP_195149.1 | 3 - 4 | 704 | 76 | ||
| GenBank mRNA | AY034919.1 | - | 3 - 4 | 483 | 76 | |
| AY113919.1 | - | 3 - 4 | 430 | 76 | ||
| AY302082.1 | - | 3 - 4 | 430 | 76 | ||
More Information
Set Membership: |
|

CAD5
Genomic Map