Assay Details
| Gene Symbol: XLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342282.1 | NP_001328987.1 | 3 - 4 | 1744 | 80 | |
| NM_119604.4 | NP_195165.2 | 3 - 4 | 1767 | 80 | ||
| GenBank mRNA | GQ229389.1 | - | 3 - 4 | 1417 | 80 | |
More Information
Set Membership: |
|

XLG2
Genomic Map