Assay Details
| Gene Symbol: XLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342282.1 | NP_001328987.1 | 5 - 6 | 2057 | 93 | |
| NM_119604.4 | NP_195165.2 | 5 - 6 | 2080 | 93 | ||
| GenBank mRNA | GQ229389.1 | - | 5 - 6 | 1730 | 93 | |
More Information
Set Membership: |
|

XLG2
Genomic Map