Assay Details
| Gene Symbol: COW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342293.1 | NP_001328041.1 | 3 - 4 | 414 | 100 | |
| NM_119624.3 | NP_195184.2 | 2 - 3 | 302 | 100 | ||
More Information
Set Membership: |
|

COW1
Genomic Map