Assay Details
| Gene Symbol: ASE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119640.3 | NP_195200.1 | 1 | 2027 | 150 | |
| GenBank mRNA | AY065123.1 | - | 1 | 1945 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASE2| Gene Symbol: ASE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119640.3 | NP_195200.1 | 1 | 2027 | 150 | |
| GenBank mRNA | AY065123.1 | - | 1 | 1945 | 150 | |
Set Membership: |
|