Assay Details
| Gene Symbol: LAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342307.1 | NP_001328609.1 | 1 - 2 | 898 | 83 | |
| NM_119651.4 | NP_567971.1 | 2 - 3 | 903 | 83 | ||
| GenBank mRNA | AY085918.1 | - | 2 - 3 | 794 | 83 | |
| DQ062353.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062354.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062355.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062356.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062357.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062359.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062360.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062362.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062365.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062366.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062369.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062370.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062371.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062372.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062373.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062374.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062375.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062377.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062378.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062379.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062381.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062382.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062383.1 | - | 2 - 3 | 758 | 83 | ||
| DQ062384.1 | - | 2 - 3 | 758 | 83 | ||
| DQ446894.1 | - | 2 - 3 | 758 | 83 | ||
More Information
Set Membership: |
|

LAP5
Genomic Map