Assay Details
| Gene Symbol: XDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119655.4 | NP_195215.2 | 11 - 12 | 3642 | 119 | |
| GenBank mRNA | AY171562.1 | - | 11 - 12 | 3439 | 119 | |
More Information
Set Membership: |
|

XDH1
Genomic Map