Assay Details
| Gene Symbol: XDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119655.4 | NP_195215.2 | 1 - 2 | 328 | 70 | |
| GenBank mRNA | AY171562.1 | - | 1 - 2 | 125 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XDH1| Gene Symbol: XDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119655.4 | NP_195215.2 | 1 - 2 | 328 | 70 | |
| GenBank mRNA | AY171562.1 | - | 1 - 2 | 125 | 70 | |
Set Membership: |
|