Assay Details
| Gene Symbol: AT4G34910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119657.3 | NP_195217.1 | 11 - 12 | 1675 | 72 | |
| GenBank mRNA | BT002816.1 | - | 11 - 12 | 1582 | 72 | |
| BT004442.1 | - | 11 - 12 | 1454 | 72 | ||
More Information
Set Membership: |
|

AT4G34910
Genomic Map