Assay Details
| Gene Symbol: AT4G34910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119657.3 | NP_195217.1 | 2 - 3 | 622 | 83 | |
| GenBank mRNA | BT002816.1 | - | 2 - 3 | 529 | 83 | |
| BT004442.1 | - | 2 - 3 | 401 | 83 | ||
More Information
Set Membership: |
|

AT4G34910
Genomic Map