Assay Details
| Gene Symbol: FAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198471.1 | NP_001185400.1 | 2 - 3 | 570 | 74 | |
| NM_106154.4 | NP_565097.1 | 1 - 2 | 720 | 74 | ||
| NM_179557.4 | NP_849888.1 | 1 - 2 | 720 | 74 | ||
| GenBank mRNA | AF318307.1 | - | 1 - 2 | 405 | 74 | |
| AF419598.1 | - | 1 - 2 | 484 | 74 | ||
| AY054196.1 | - | 1 - 2 | 565 | 74 | ||
| AY097344.1 | - | 1 - 2 | 318 | 74 | ||
More Information
Set Membership: |
|

FAB1
Genomic Map