Assay Details
| Gene Symbol: APL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106591.2 | NP_565209.1 | 4 - 5 | 592 | 63 | |
| NM_179574.3 | NP_849905.1 | 5 - 6 | 577 | 63 | ||
| GenBank mRNA | AK316719.1 | - | 5 - 6 | 503 | 63 | |
| AY045662.1 | - | 4 - 5 | 590 | 63 | ||
| AY143841.1 | - | 4 - 5 | 238 | 63 | ||
More Information
Set Membership: |
|

APL
Genomic Map