Assay Details
| Gene Symbol: AT1G80030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334948.1 | NP_001321726.1 | 1 - 2 | 390 | 96 | |
| NM_106652.3 | NP_565227.1 | 1 - 2 | 511 | 96 | ||
| NM_179579.4 | NP_849910.1 | 1 - 2 | 511 | 96 | ||
| NM_179580.2 | NP_849911.1 | 1 - 2 | 511 | 96 | ||
| GenBank mRNA | AK229599.1 | - | 1 - 2 | 385 | 96 | |
More Information
Set Membership: |
|

AT1G80030
Genomic Map