Assay Details
| Gene Symbol: AT1G80620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179583.5 | NP_849914.1 | 2 - 3 | 1321 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G80620| Gene Symbol: AT1G80620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179583.5 | NP_849914.1 | 2 - 3 | 1321 | 89 | |
Set Membership: |
|