Assay Details
| Gene Symbol: CALS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179622.4 | NP_849953.2 | 3 - 4 | 363 | 66 | |
| GenBank mRNA | AY337762.1 | - | 3 - 4 | 346 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALS5| Gene Symbol: CALS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179622.4 | NP_849953.2 | 3 - 4 | 363 | 66 | |
| GenBank mRNA | AY337762.1 | - | 3 - 4 | 346 | 66 | |
Set Membership: |
|