Assay Details
| Gene Symbol: CALS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179622.4 | NP_849953.2 | 24 - 25 | 2653 | 108 | |
| GenBank mRNA | AY337762.1 | - | 24 - 25 | 2636 | 108 | |
More Information
Set Membership: |
|

CALS5
Genomic Map