Assay Details
| Gene Symbol: CALS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179622.4 | NP_849953.2 | 38 - 39 | 4316 | 144 | |
| GenBank mRNA | AY337762.1 | - | 38 - 39 | 4299 | 144 | |
More Information
Set Membership: |
|

CALS5
Genomic Map