Assay Details
| Gene Symbol: DGK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179649.4 | NP_849980.1 | 2 - 3 | 727 | 64 | |
| GenBank mRNA | AY064982.1 | - | 2 - 3 | 576 | 64 | |
| AY141990.1 | - | 2 - 3 | 487 | 64 | ||
More Information
Set Membership: |
|

DGK3
Genomic Map