Assay Details
| Gene Symbol: LKP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179652.2 | NP_849983.1 | 1 - 2 | 473 | 126 | |
| GenBank mRNA | AB038797.1 | - | 1 - 2 | 474 | 126 | |
| AY099709.1 | - | 1 - 2 | 433 | 126 | ||
| BT046116.1 | - | 1 - 2 | 245 | 126 | ||
More Information
Set Membership: |
|

LKP2
Genomic Map