Assay Details
| Gene Symbol: DWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179658.4 | NP_849989.1 | 3 - 4 | 622 | 68 | |
| GenBank mRNA | AY099718.1 | - | 3 - 4 | 475 | 68 | |
| AY128879.1 | - | 3 - 4 | 451 | 68 | ||
More Information
Set Membership: |
|

DWA1
Genomic Map