Assay Details
| Gene Symbol: SR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124889.2 | NP_001118361.1 | 2 - 3 | 340 | 71 | |
| NM_179692.3 | NP_850023.1 | 1 - 2 | 412 | 71 | ||
| GenBank mRNA | AF506697.1 | - | 1 - 2 | 36 | 71 | |
| AY510025.1 | - | 1 - 2 | 36 | 71 | ||
| BT002459.1 | - | 1 - 2 | 219 | 71 | ||
More Information
Set Membership: |
|

SR1
Genomic Map