Assay Details
| Gene Symbol: AGL61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179727.5 | NP_850058.1 | 1 | 752 | 86 | |
| GenBank mRNA | DQ056542.1 | - | 1 | 739 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL61| Gene Symbol: AGL61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179727.5 | NP_850058.1 | 1 | 752 | 86 | |
| GenBank mRNA | DQ056542.1 | - | 1 | 739 | 86 | |
Set Membership: |
|