Assay Details
| Gene Symbol: HCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336011.1 | NP_001324826.1 | 7 - 8 | 1498 | 102 | |
| NM_001336012.1 | NP_001324829.1 | 8 - 9 | 1118 | 102 | ||
| NM_001336013.1 | NP_001324830.1 | 9 - 10 | 999 | 102 | ||
| NM_001336015.1 | NP_001324828.1 | 6 - 7 | 1623 | 102 | ||
| NM_001336016.1 | NP_001324831.1 | 8 - 9 | 1383 | 102 | ||
| NM_128130.3 | NP_565605.1 | 8 - 9 | 1162 | 102 | ||
| NM_179736.1 | NP_850067.1 | 9 - 10 | 1047 | 102 | ||
| GenBank mRNA | BT024748.1 | - | 8 - 9 | 990 | 102 | |
| U41369.1 | - | 8 - 9 | 1039 | 102 | ||
More Information
Set Membership: |
|

HCS1
Genomic Map