Assay Details
| Gene Symbol: PAPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036340.1 | NP_001031417.1 | 8 - 9 | 1241 | 90 | |
| NM_001202674.2 | NP_001189603.1 | 8 - 9 | 1390 | 90 | ||
| NM_001336022.1 | NP_001324471.1 | 8 - 9 | 1422 | 90 | ||
| NM_001336023.1 | NP_001324473.1 | 8 - 9 | 1422 | 90 | ||
| NM_001336024.1 | NP_001324472.1 | 8 - 9 | 1422 | 90 | ||
| NM_001336025.1 | NP_001324470.1 | 8 - 9 | 1411 | 90 | ||
| NM_001336026.1 | NP_001324469.1 | 8 - 9 | 1375 | 90 | ||
| NM_128145.4 | NP_565611.1 | 8 - 9 | 1422 | 90 | ||
| NM_179740.2 | NP_850071.1 | 8 - 9 | 1422 | 90 | ||
| GenBank mRNA | AF255297.2 | - | 8 - 9 | 1054 | 90 | |
| AY039974.1 | - | 8 - 9 | 1242 | 90 | ||
| AY074533.1 | - | 8 - 9 | 1054 | 90 | ||
More Information
Set Membership: |
|

PAPS2
Genomic Map