Assay Details
| Gene Symbol: AT2G26690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336079.1 | NP_001324098.1 | 2 - 3 | 584 | 65 | |
| NM_001336080.1 | NP_001324099.1 | 1 - 2 | 266 | 65 | ||
| NM_179753.1 | NP_850084.1 | 3 - 4 | 403 | 65 | ||
| GenBank mRNA | AJ011604.1 | - | 2 - 3 | 331 | 65 | |
More Information
Set Membership: |
|

AT2G26690
Genomic Map