Assay Details
| Gene Symbol: AT2G26690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336079.1 | NP_001324098.1 | 5 - 6 | 1470 | 94 | |
| NM_001336080.1 | NP_001324099.1 | 4 - 5 | 1152 | 94 | ||
| NM_179753.1 | NP_850084.1 | 6 - 7 | 1289 | 94 | ||
| GenBank mRNA | AJ011604.1 | - | 5 - 6 | 1217 | 94 | |
More Information
Set Membership: |
|

AT2G26690
Genomic Map