Assay Details
| Gene Symbol: NLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179770.1 | NP_850101.1 | 1 - 2 | 192 | 145 | |
| GenBank mRNA | AY072113.1 | - | 1 - 2 | 192 | 145 | |
| AY122963.1 | - | 1 - 2 | 171 | 145 | ||
More Information
Set Membership: |
|

NLP1
Genomic Map