Assay Details
| Gene Symbol: HDT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336129.1 | NP_001318300.1 | 4 - 5 | 415 | 228 | |
| NM_128344.4 | NP_565661.2 | 4 - 5 | 603 | 135 | ||
| NM_179778.1 | NP_850109.1 | 3 - 4 | 325 | 135 | ||
| GenBank mRNA | AF255713.1 | - | 3 - 4 | 324 | 135 | |
| AK229619.1 | - | 4 - 5 | 388 | 135 | ||
| BT006162.1 | - | 4 - 5 | 388 | 135 | ||
| BT008535.1 | - | 3 - 4 | 324 | 135 | ||
More Information
Set Membership: |
|

HDT4
Genomic Map