Assay Details
| Gene Symbol: HDT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336129.1 | NP_001318300.1 | 5 - 6 | 610 | 115 | |
| NM_128344.4 | NP_565661.2 | 6 - 7 | 705 | 115 | ||
| NM_179778.1 | NP_850109.1 | 5 - 6 | 427 | 115 | ||
| GenBank mRNA | AF255713.1 | - | 5 - 6 | 426 | 115 | |
| AK229619.1 | - | 6 - 7 | 490 | 115 | ||
| BT006162.1 | - | 6 - 7 | 490 | 115 | ||
| BT008535.1 | - | 5 - 6 | 426 | 115 | ||
More Information
Set Membership: |
|

HDT4
Genomic Map