Assay Details
| Gene Symbol: HDT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336129.1 | NP_001318300.1 | 6 - 7 | 745 | 131 | |
| NM_001336130.1 | NP_001324791.1 | 6 - 7 | 716 | 131 | ||
| NM_128344.4 | NP_565661.2 | 7 - 8 | 840 | 131 | ||
| NM_179778.1 | NP_850109.1 | 6 - 7 | 562 | 131 | ||
| GenBank mRNA | AF255713.1 | - | 6 - 7 | 561 | 131 | |
| AK229619.1 | - | 7 - 8 | 625 | 131 | ||
| BT006162.1 | - | 7 - 8 | 625 | 131 | ||
| BT008535.1 | - | 6 - 7 | 561 | 131 | ||
More Information
Set Membership: |
|

HDT4
Genomic Map