Assay Details
| Gene Symbol: ABCG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336143.1 | NP_001318302.1 | 3 - 4 | 775 | 129 | |
| NM_179780.1 | NP_850111.1 | 2 - 3 | 571 | 129 | ||
More Information
Set Membership: |
|

ABCG3
Genomic Map