Assay Details
| Gene Symbol: ABCG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336143.1 | NP_001318302.1 | 10 - 11 | 2040 | 74 | |
| NM_179780.1 | NP_850111.1 | 9 - 10 | 1836 | 74 | ||
More Information
Set Membership: |
|

ABCG3
Genomic Map