Assay Details
| Gene Symbol: FRA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179782.4 | NP_850113.2 | 1 - 2 | 626 | 148 | |
| GenBank mRNA | DQ182567.1 | - | 1 - 2 | 305 | 148 | |
| KJ138894.1 | - | 1 - 2 | 264 | 148 | ||
More Information
Set Membership: |
|

FRA8
Genomic Map