Assay Details
| Gene Symbol: FRU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336147.1 | NP_001323907.1 | 1 - 2 | 495 | 100 | |
| NM_179783.2 | NP_850114.1 | 1 - 2 | 537 | 100 | ||
| GenBank mRNA | AB493568.1 | - | 1 - 2 | 482 | 100 | |
| BT026446.1 | - | 1 - 2 | 482 | 100 | ||
More Information
Set Membership: |
|

FRU
Genomic Map