Assay Details
| Gene Symbol: FRU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336147.1 | NP_001323907.1 | 3 - 4 | 874 | 75 | |
| NM_179783.2 | NP_850114.1 | 3 - 4 | 916 | 75 | ||
| GenBank mRNA | AB493568.1 | - | 3 - 4 | 861 | 75 | |
| BT026446.1 | - | 3 - 4 | 861 | 75 | ||
More Information
Set Membership: |
|

FRU
Genomic Map