Assay Details
| Gene Symbol: BGAL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202695.2 | NP_001189624.1 | 12 - 13 | 1677 | 84 | |
| NM_001336165.1 | NP_001324192.1 | 10 - 11 | 1928 | 84 | ||
| NM_001336166.1 | NP_001324193.1 | 11 - 12 | 1781 | 84 | ||
| NM_001336167.1 | NP_001324194.1 | 10 - 11 | 1616 | 84 | ||
| NM_179790.3 | NP_850121.1 | 11 - 12 | 1824 | 84 | ||
More Information
Set Membership: |
|

BGAL8
Genomic Map