Assay Details
| Gene Symbol: BGAL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202695.2 | NP_001189624.1 | 15 - 16 | 2155 | 107 | |
| NM_001336165.1 | NP_001324192.1 | 13 - 14 | 2406 | 107 | ||
| NM_001336166.1 | NP_001324193.1 | 14 - 15 | 2259 | 107 | ||
| NM_001336167.1 | NP_001324194.1 | 13 - 14 | 2094 | 107 | ||
| NM_179790.3 | NP_850121.1 | 14 - 15 | 2302 | 107 | ||
More Information
Set Membership: |
|

BGAL8
Genomic Map