Assay Details
| Gene Symbol: BGAL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202695.2 | NP_001189624.1 | 9 - 10 | 1358 | 149 | |
| NM_001336165.1 | NP_001324192.1 | 7 - 8 | 1609 | 149 | ||
| NM_001336166.1 | NP_001324193.1 | 8 - 9 | 1462 | 149 | ||
| NM_001336167.1 | NP_001324194.1 | 7 - 8 | 1297 | 149 | ||
| NM_179790.3 | NP_850121.1 | 8 - 9 | 1505 | 149 | ||
More Information
Set Membership: |
|

BGAL8
Genomic Map