Assay Details
Gene Symbol: VHA-A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179791.2 | NP_850122.1 | 2 - 3 | 411 | 89 | |
GenBank mRNA | AY091008.1 | - | 2 - 3 | 315 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
