Assay Details
| Gene Symbol: HSI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179815.3 | NP_850146.1 | 2 - 3 | 757 | 68 | |
| GenBank mRNA | AB206553.1 | - | 2 - 3 | 512 | 68 | |
| AY062492.1 | - | 2 - 3 | 520 | 68 | ||
More Information
Set Membership: |
|

HSI2
Genomic Map