Assay Details
| Gene Symbol: HSI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179815.3 | NP_850146.1 | 8 - 9 | 2117 | 94 | |
| GenBank mRNA | AB206553.1 | - | 8 - 9 | 1872 | 94 | |
| AY062492.1 | - | 8 - 9 | 1880 | 94 | ||
More Information
Set Membership: |
|

HSI2
Genomic Map