Assay Details
| Gene Symbol: LGT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179819.2 | NP_850150.1 | 5 - 6 | 715 | 103 | |
| GenBank mRNA | AY081313.1 | - | 5 - 6 | 708 | 103 | |
| BT002563.1 | - | 5 - 6 | 403 | 103 | ||
| KJ138886.1 | - | 5 - 6 | 403 | 103 | ||
More Information
Set Membership: |
|

LGT5
Genomic Map