Assay Details
| Gene Symbol: ERF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179831.2 | NP_850162.1 | 1 | 853 | 115 | |
| GenBank mRNA | AY070443.1 | - | 1 | 755 | 115 | |
| AY096563.1 | - | 1 | 688 | 115 | ||
More Information
Set Membership: |
|

ERF15
Genomic Map