Assay Details
| Gene Symbol: PARG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336350.1 | NP_001324969.1 | 7 - 8 | 1208 | 70 | |
| NM_179844.3 | NP_850175.1 | 7 - 8 | 1208 | 70 | ||
| NM_201849.3 | NP_973578.1 | 7 - 8 | 1208 | 70 | ||
| GenBank mRNA | AY072330.1 | - | 7 - 8 | 1173 | 70 | |
| BT010379.1 | - | 7 - 8 | 1109 | 70 | ||
More Information
Set Membership: |
|

PARG2
Genomic Map